Assay Details
| Gene Symbol: pcdh8.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004845.1 | NP_001004845.1 | 1 - 2 | 2118 | 120 | |
| GenBank mRNA | BC074660.1 | - | 1 - 2 | 2118 | 120 | |
More Information
Set Membership: |
|

pcdh8.2
Genomic Map