Assay Details
| Gene Symbol: otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004849.1 | NP_001004849.1 | 1486 | 68 | ||
| GenBank mRNA | BC074667.1 | - | 1486 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

otud5| Gene Symbol: otud5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004849.1 | NP_001004849.1 | 1486 | 68 | ||
| GenBank mRNA | BC074667.1 | - | 1486 | 68 | ||
Set Membership: |
|