Assay Details
| Gene Symbol: abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004867.1 | NP_001004867.1 | 2 - 3 | 845 | 98 | |
| GenBank mRNA | BC074709.1 | - | 2 - 3 | 845 | 98 | |
More Information
Set Membership: |
|

abhd17c
Genomic Map