Assay Details
| Gene Symbol: n4bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004870.1 | NP_001004870.1 | 4 - 5 | 1865 | 66 | |
| XM_012960812.2 | XP_012816266.1 | 4 - 5 | 2015 | 66 | ||
| GenBank mRNA | BC075104.1 | - | 4 - 5 | 1865 | 66 | |
| CT025272.2 | - | 4 - 5 | 1841 | 66 | ||
More Information
Set Membership: |
|

n4bp1
Genomic Map