Assay Details
| Gene Symbol: tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004894.1 | NP_001004894.1 | 3 - 4 | 400 | 87 | |
| XM_018092022.1 | XP_017947511.1 | 3 - 4 | 507 | 87 | ||
| GenBank mRNA | BC075302.1 | - | 3 - 4 | 400 | 87 | |
| CR761221.2 | - | 3 - 4 | 394 | 87 | ||
More Information
Set Membership: |
|

tnfrsf10b
Genomic Map