Assay Details
| Gene Symbol: tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004894.1 | NP_001004894.1 | 5 - 6 | 582 | 74 | |
| XM_018092022.1 | XP_017947511.1 | 5 - 6 | 689 | 74 | ||
| GenBank mRNA | BC075302.1 | - | 5 - 6 | 582 | 74 | |
| CR761221.2 | - | 5 - 6 | 576 | 74 | ||
More Information
Set Membership: |
|

tnfrsf10b
Genomic Map