Assay Details
| Gene Symbol: tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004894.1 | NP_001004894.1 | 7 - 8 | 695 | 81 | |
| GenBank mRNA | BC075302.1 | - | 7 - 8 | 695 | 81 | |
| CR761221.2 | - | 7 - 8 | 689 | 81 | ||
More Information
Set Membership: |
|

tnfrsf10b
Genomic Map