Assay Details
| Gene Symbol: gapdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004949.2 | NP_001004949.1 | 5 - 6 | 466 | 94 | |
| XM_012966386.2 | XP_012821840.1 | 6 - 7 | 680 | 94 | ||
| GenBank mRNA | BC075438.1 | - | 5 - 6 | 467 | 94 | |
| CR760856.2 | - | 5 - 6 | 482 | 94 | ||
More Information
Set Membership: |
|

gapdh
Genomic Map