Assay Details
| Gene Symbol: aadac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004960.1 | NP_001004960.1 | 2 - 3 | 475 | 69 | |
| GenBank mRNA | BC075468.1 | - | 2 - 3 | 475 | 69 | |
More Information
Set Membership: |
|

aadac
Genomic Map