Assay Details
| Gene Symbol: washc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004970.1 | NP_001004970.1 | 4 - 5 | 572 | 65 | |
| GenBank mRNA | BC075486.1 | - | 4 - 5 | 572 | 65 | |
More Information
Set Membership: |
|

washc5
Genomic Map