Assay Details
| Gene Symbol: rpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005026.1 | NP_001005026.1 | 3 - 4 | 266 | 63 | |
| XM_018091168.1 | XP_017946657.1 | 4 - 5 | 323 | 63 | ||
| GenBank mRNA | BC076707.1 | - | 3 - 4 | 266 | 63 | |
| BC157264.1 | - | 1502 | 63 | |||
| CT030557.1 | - | 3 - 4 | 264 | 63 | ||
More Information
Set Membership: |
|

rpl21
Genomic Map