Assay Details
| Gene Symbol: btg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011013.1 | NP_001011013.1 | 1 - 2 | 225 | 72 | |
| GenBank mRNA | BC082726.1 | - | 1 - 2 | 225 | 72 | |
| CR761962.2 | - | 1 - 2 | 238 | 72 | ||
More Information
Set Membership: |
|

btg2
Genomic Map