Assay Details
| Gene Symbol: gcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011024.1 | NP_001011024.1 | 3 - 4 | 289 | 79 | |
| GenBank mRNA | BC082740.1 | - | 3 - 4 | 289 | 79 | |
| CR761928.2 | - | 3 - 4 | 287 | 79 | ||
More Information
Set Membership: |
|

gcsh
Genomic Map