Assay Details
| Gene Symbol: oaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300762.1 | NP_001287691.1 | 1 - 2 | 249 | 76 | |
| NM_203549.2 | NP_988880.3 | 1 - 2 | 249 | 76 | ||
| GenBank mRNA | AL641155.2 | - | 1 - 2 | 249 | 76 | |
| BC059754.1 | - | 1 - 2 | 171 | 76 | ||
| CX741301.1 | - | 1 - 2 | 182 | 76 | ||
| DR838823.1 | - | 4 - 5 | 721 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059754.1 transcripts.
Set Membership: |
|

oaz1
Genomic Map