Assay Details
| Gene Symbol: rpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203550.1 | NP_988881.1 | 4 - 5 | 389 | 85 | |
| GenBank mRNA | BC059751.1 | - | 4 - 5 | 389 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl5| Gene Symbol: rpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203550.1 | NP_988881.1 | 4 - 5 | 389 | 85 | |
| GenBank mRNA | BC059751.1 | - | 4 - 5 | 389 | 85 | |
Set Membership: |
|