Assay Details
| Gene Symbol: clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203558.1 | NP_988889.1 | 5 - 6 | 607 | 70 | |
| XM_012968147.2 | XP_012823601.1 | 5 - 6 | 588 | 70 | ||
| GenBank mRNA | BC059765.1 | - | 5 - 6 | 607 | 70 | |
| CR855678.2 | - | 5 - 6 | 672 | 70 | ||
| CR942528.2 | - | 5 - 6 | 596 | 70 | ||
More Information
Set Membership: |
|

clic1
Genomic Map