Assay Details
| Gene Symbol: ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203644.1 | NP_988975.1 | 8 - 9 | 1151 | 68 | |
| GenBank mRNA | BC061374.1 | - | 8 - 9 | 1151 | 68 | |
More Information
Set Membership: |
|

ap2m1
Genomic Map