Assay Details
| Gene Symbol: tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203665.1 | NP_988996.1 | 1 - 2 | 175 | 89 | |
| GenBank mRNA | BC061269.1 | - | 1 - 2 | 175 | 89 | |
| BC122046.1 | - | 1 - 2 | 163 | 89 | ||
| CU075322.1 | - | 1 - 2 | 143 | 89 | ||
More Information
Set Membership: |
|

tmem38a
Genomic Map