Assay Details
| Gene Symbol: mob2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203682.1 | NP_989013.1 | 2 - 3 | 247 | 89 | |
| XM_012960726.2 | XP_012816180.1 | 2 - 3 | 434 | 89 | ||
| XM_012960727.2 | XP_012816181.2 | 2 - 3 | 270 | 89 | ||
| XM_012960729.2 | XP_012816183.1 | 2 - 3 | 514 | 89 | ||
| GenBank mRNA | BC061432.1 | - | 2 - 3 | 247 | 89 | |
More Information
Set Membership: |
|

mob2.1
Genomic Map