Assay Details
| Gene Symbol: fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203721.1 | NP_989052.1 | 7 - 8 | 945 | 94 | |
| XM_018091140.1 | XP_017946629.1 | 7 - 8 | 970 | 94 | ||
| GenBank mRNA | BC061340.1 | - | 7 - 8 | 945 | 94 | |
More Information
Set Membership: |
|

fkbp6
Genomic Map