Assay Details
| Gene Symbol: psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203740.1 | NP_989071.1 | 3 - 4 | 410 | 87 | |
| GenBank mRNA | BC061282.1 | - | 3 - 4 | 410 | 87 | |
| CR760155.2 | - | 3 - 4 | 385 | 87 | ||
More Information
Set Membership: |
|

psma7
Genomic Map