Assay Details
| Gene Symbol: psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203740.1 | NP_989071.1 | 5 - 6 | 646 | 94 | |
| GenBank mRNA | BC061282.1 | - | 5 - 6 | 646 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

psma7| Gene Symbol: psma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203740.1 | NP_989071.1 | 5 - 6 | 646 | 94 | |
| GenBank mRNA | BC061282.1 | - | 5 - 6 | 646 | 94 | |
Set Membership: |
|