Assay Details
| Gene Symbol: act3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC061264.1 | - | 2 - 3 | 180 | 72 | |
| CR760277.2 | - | 1 - 2 | 158 | 72 | ||
| CT025518.2 | - | 979 | 72 | |||
More Information
Set Membership: |
|

act3
Genomic Map