Assay Details
| Gene Symbol: atp5pb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203787.1 | NP_989118.1 | 3 - 4 | 244 | 61 | |
| GenBank mRNA | BC061296.1 | - | 3 - 4 | 244 | 61 | |
| CR760878.2 | - | 3 - 4 | 276 | 61 | ||
More Information
Set Membership: |
|

atp5pb
Genomic Map