Assay Details
| Gene Symbol: rab3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203799.1 | NP_989130.1 | 5 - 6 | 711 | 72 | |
| XM_012958764.1 | XP_012814218.1 | 5 - 6 | 809 | 72 | ||
| XM_012958765.2 | XP_012814219.1 | 5 - 6 | 834 | 72 | ||
| XM_012958766.2 | XP_012814220.1 | 5 - 6 | 726 | 72 | ||
| XM_018092001.1 | XP_017947490.1 | 5 - 6 | 824 | 72 | ||
| XM_018092003.1 | XP_017947492.1 | 5 - 6 | 821 | 72 | ||
| XM_018092004.1 | XP_017947493.1 | 5 - 6 | 738 | 72 | ||
| GenBank mRNA | BC061285.1 | - | 5 - 6 | 711 | 72 | |
| CR761650.2 | - | 4 - 5 | 677 | 72 | ||
More Information
Set Membership: |
|

rab3ip
Genomic Map