Assay Details
| Gene Symbol: prps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203809.1 | NP_989140.1 | 6 - 7 | 913 | 69 | |
| XM_012968153.2 | XP_012823607.1 | 6 - 7 | 773 | 69 | ||
| GenBank mRNA | BC061401.1 | - | 6 - 7 | 913 | 69 | |
More Information
Set Membership: |
|

prps1
Genomic Map