Assay Details
| Gene Symbol: ywhaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203842.1 | NP_989173.1 | 2 - 3 | 677 | 69 | |
| XM_018094585.1 | XP_017950074.1 | 2 - 3 | 486 | 69 | ||
| GenBank mRNA | BC063188.1 | - | 2 - 3 | 677 | 69 | |
| CR926430.2 | - | 1 - 2 | 317 | 69 | ||
More Information
Set Membership: |
|

ywhaz
Genomic Map