Assay Details
| Gene Symbol: tktl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203896.1 | NP_989227.1 | 8 - 9 | 1158 | 66 | |
| GenBank mRNA | BC063226.1 | - | 8 - 9 | 1158 | 66 | |
More Information
Set Membership: |
|

tktl2
Genomic Map