Assay Details
| Gene Symbol: gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203919.1 | NP_989250.1 | 4 - 5 | 661 | 65 | |
| XM_012964457.2 | XP_012819911.2 | 3 - 4 | 349 | 65 | ||
| GenBank mRNA | BC063931.1 | - | 4 - 5 | 661 | 65 | |
More Information
Set Membership: |
|

gnai2
Genomic Map