Assay Details
| Gene Symbol: gyg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203939.1 | NP_989270.1 | 5 - 6 | 694 | 65 | |
| GenBank mRNA | BC064206.1 | - | 5 - 6 | 694 | 65 | |
| CR760381.2 | - | 5 - 6 | 671 | 65 | ||
More Information
Set Membership: |
|

gyg1
Genomic Map