Assay Details
| Gene Symbol: arglu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203949.1 | NP_989280.1 | 2 - 3 | 626 | 70 | |
| GenBank mRNA | BC064271.1 | - | 2 - 3 | 626 | 70 | |
More Information
Set Membership: |
|

arglu1
Genomic Map