Assay Details
| Gene Symbol: glul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203966.1 | NP_989297.1 | 6 - 7 | 866 | 70 | |
| XM_012969756.1 | XP_012825210.1 | 6 - 7 | 924 | 70 | ||
| GenBank mRNA | BC064185.1 | - | 6 - 7 | 866 | 70 | |
| CR926251.1 | - | 6 - 7 | 844 | 70 | ||
More Information
Set Membership: |
|

glul
Genomic Map