Assay Details
| Gene Symbol: srsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203997.1 | NP_989328.1 | 2 - 3 | 790 | 61 | |
| GenBank mRNA | BC064167.1 | - | 2 - 3 | 790 | 61 | |
| CR760874.2 | - | 2 - 3 | 698 | 61 | ||
More Information
Set Membership: |
|

srsf2
Genomic Map