Assay Details
| Gene Symbol: ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204044.1 | NP_989375.1 | 3 - 4 | 437 | 107 | |
| GenBank mRNA | BC064184.1 | - | 3 - 4 | 437 | 107 | |
| CR762229.2 | - | 3 - 4 | 440 | 107 | ||
More Information
Set Membership: |
|

ube2n
Genomic Map