Assay Details
| Gene Symbol: coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204052.1 | NP_989383.1 | 2 - 3 | 332 | 66 | |
| XM_012958577.2 | XP_012814031.1 | 2 - 3 | 226 | 66 | ||
| XM_018091891.1 | XP_017947380.1 | 2 - 3 | 434 | 66 | ||
| GenBank mRNA | BC064872.1 | - | 2 - 3 | 332 | 66 | |
| BC080158.1 | - | 2 - 3 | 365 | 66 | ||
More Information
Set Membership: |
|

coro1c
Genomic Map