Assay Details
| Gene Symbol: cdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204086.1 | NP_989417.1 | 2 - 3 | 716 | 64 | |
| GenBank mRNA | AF417199.1 | - | 2 - 3 | 716 | 64 | |
| BC167866.1 | - | 2 - 3 | 718 | 64 | ||
| BC170865.1 | - | 2 - 3 | 606 | 64 | ||
| BC170869.1 | - | 2 - 3 | 606 | 64 | ||
| CT030483.1 | - | 2 - 3 | 698 | 64 | ||
More Information
Set Membership: |
|

cdx4
Genomic Map