Assay Details
| Gene Symbol: snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204093.1 | NP_989424.1 | 1 - 2 | 226 | 90 | |
| GenBank mRNA | AF368039.1 | - | 1 - 2 | 226 | 90 | |
| BC075376.1 | - | 1 - 2 | 185 | 90 | ||
| CR926188.2 | - | 1 - 2 | 207 | 90 | ||
More Information
Set Membership: |
|

snai2
Genomic Map