Assay Details
| Gene Symbol: pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205830.1 | NP_991399.1 | 9 - 10 | 1475 | 60 | |
| GenBank mRNA | BC066132.1 | - | 9 - 10 | 1475 | 60 | |
| CR760201.2 | - | 9 - 10 | 1473 | 60 | ||
More Information
Set Membership: |
|

pafah1b1
Genomic Map