Assay Details
| Gene Symbol: ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213678.1 | NP_998843.1 | 5 - 6 | 704 | 84 | |
| XM_012966324.2 | XP_012821778.1 | 5 - 6 | 464 | 84 | ||
| XM_012966326.2 | XP_012821780.1 | 5 - 6 | 464 | 84 | ||
| XM_012966327.2 | XP_012821781.1 | 5 - 6 | 956 | 84 | ||
| XM_012966328.2 | XP_012821782.1 | 5 - 6 | 381 | 84 | ||
| XM_012966329.2 | XP_012821783.1 | 5 - 6 | 464 | 84 | ||
| XM_012966330.2 | XP_012821784.1 | 5 - 6 | 464 | 84 | ||
| GenBank mRNA | BC067989.1 | - | 5 - 6 | 704 | 84 | |
More Information
Set Membership: |
|

ldb1
Genomic Map