Assay Details
| Gene Symbol: cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC067328.1 | - | 3 - 4 | 499 | 70 | |
| BC080356.1 | - | 3 - 4 | 425 | 70 | ||
| CR760270.2 | - | 2 - 3 | 392 | 70 | ||
More Information
Set Membership: |
|

cfl1
Genomic Map