Assay Details
| Gene Symbol: rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005122.2 | NP_001005122.1 | 4 - 5 | 383 | 87 | |
| GenBank mRNA | BC077657.1 | - | 4 - 5 | 384 | 87 | |
More Information
Set Membership: |
|

rpl19
Genomic Map