Assay Details
| Gene Symbol: dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005145.2 | NP_001005145.1 | 5 - 6 | 362 | 62 | |
| GenBank mRNA | BC077694.1 | - | 5 - 6 | 365 | 62 | |
More Information
Set Membership: |
|

dnajc15
Genomic Map