Assay Details
| Gene Symbol: sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005146.2 | NP_001005146.2 | 4 - 5 | 507 | 71 | |
| GenBank mRNA | BC077695.1 | - | 3 - 4 | 392 | 71 | |
| CT025343.2 | - | 2 - 3 | 139 | 71 | ||
| CU025143.1 | - | 4 - 5 | 479 | 71 | ||
| EL673915.1 | - | 4 - 5 | 507 | 71 | ||
More Information
Set Membership: |
|

sft2d1
Genomic Map