Assay Details
| Gene Symbol: elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005461.1 | NP_001005461.1 | 2 - 3 | 241 | 100 | |
| GenBank mRNA | BC074585.1 | - | 2 - 3 | 241 | 100 | |
| CT030443.1 | - | 1 - 2 | 188 | 100 | ||
More Information
Set Membership: |
|

elavl1
Genomic Map