Assay Details
| Gene Symbol: elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005461.1 | NP_001005461.1 | 4 - 5 | 496 | 91 | |
| GenBank mRNA | BC074585.1 | - | 4 - 5 | 496 | 91 | |
| CT030443.1 | - | 3 - 4 | 443 | 91 | ||
More Information
Set Membership: |
|

elavl1
Genomic Map