Assay Details
| Gene Symbol: epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005627.1 | NP_001005627.1 | 4 - 5 | 701 | 86 | |
| XM_018092016.1 | XP_017947505.1 | 5 - 6 | 822 | 86 | ||
| GenBank mRNA | BC074612.1 | - | 4 - 5 | 701 | 86 | |
More Information
Set Membership: |
|

epyc
Genomic Map