Assay Details
| Gene Symbol: tcf7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005640.1 | NP_001005640.1 | 2 - 3 | 500 | 70 | |
| GenBank mRNA | BC074638.1 | - | 2 - 3 | 500 | 70 | |
| CR942567.2 | - | 2 - 3 | 390 | 70 | ||
More Information
Set Membership: |
|

tcf7l1
Genomic Map