Assay Details
| Gene Symbol: mpst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005646.1 | NP_001005646.1 | 2 - 3 | 637 | 72 | |
| XM_012960804.2 | XP_012816258.1 | 2 - 3 | 723 | 72 | ||
| XM_012960806.2 | XP_012816260.1 | 2 - 3 | 728 | 72 | ||
| GenBank mRNA | BC074647.1 | - | 2 - 3 | 637 | 72 | |
| CR760267.2 | - | 1 - 2 | 619 | 72 | ||
More Information
Set Membership: |
|

mpst
Genomic Map