Assay Details
| Gene Symbol: ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005662.1 | NP_001005662.1 | 3 - 4 | 326 | 90 | |
| XM_018091970.1 | XP_017947459.1 | 2 - 3 | 363 | 90 | ||
| XM_018091973.1 | XP_017947462.1 | 3 - 4 | 239 | 90 | ||
| GenBank mRNA | BC074688.1 | - | 3 - 4 | 326 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018091970.1, XM_018091973.1 transcripts.
Set Membership: |
|

ube2k
Genomic Map