Assay Details
| Gene Symbol: rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005667.1 | NP_001005667.1 | 1 - 2 | 394 | 64 | |
| GenBank mRNA | BC074696.1 | - | 1 - 2 | 394 | 64 | |
More Information
Set Membership: |
|

rapsn
Genomic Map